Learn More
Biomatik Corporation Rat Lactoferrin (LTF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05541
Encompass_Preferred
Rat Lactoferrin (LTF) ELISA Kit for detecting LTF in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.84ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZAB1. Gene ID: 301034. Target Synonyms: HLF2; LF; GIG12; Lactotransferrin; Growth-inhibiting protein 12; Talalactoferrin; Lfcin-H; Kaliocin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3099