Learn More
Biomatik Corporation Mouse carcinoembryonic antign,CEA ELISA Kit
Supplier:  Biomatik Corporation EKC41080
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q3UKK2. Target Synonyms: CD66e, CEA, DKFZp781M2392, OTTHUMP00000199034. Restriction: For Research Use Only.
Catalog No. 50-228-7819