Learn More
Biomatik Corporation Human Interleukin 29 (Il-29) Elisa Kit
Supplier:  Biomatik Corporation EKC41135
Encompass_Preferred
Human Interleukin 29 (Il-29) Elisa Kit for detecting Interleukin 29 (Interferon, Lambda 1) in serum, plasma, cell culture supernates, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8IU54; IFNL1. Target Synonyms: IFNL1; IL29; ZCYTO21Interferon lambda-1; IFN-lambda-1; Cytokine Zcyto21; Interleukin-29; IL-29. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7826