Learn More
Biomatik Corporation Human Laminin Alpha 5 (LAMa5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05557
Encompass_Preferred
The Human Laminin Alpha 5 (LAMa5) ELISA Kit allows for quantitative determination of Laminin Alpha 5 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 250-4000 pg/mL. Sensitivity: 92 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15230. Cross Activity: No significant cross-reactivity or interference between Laminin Alpha 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3102