Learn More
Biomatik Corporation Human Laminin Gamma 1 (LAMC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05561
Encompass_Preferred
The Human Laminin Gamma 1 (LAMC1) ELISA Kit allows for quantitative determination of Laminin Gamma 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11047. Cross Activity: No significant cross-reactivity or interference between Laminin Gamma 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3103