Learn More
Biomatik Corporation Human Platelet-Derived Growth Factor Aa (Pdgf-Aa) Elisa Kit
Supplier:  Biomatik Corporation EKC42347
Encompass_Preferred
Human Platelet-Derived Growth Factor Aa (Pdgf-Aa) Elisa Kit for detecting Platelet-Derived Growth Factor Aa(Pdgf-Aa) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7875