Learn More
Biomatik Corporation Human ICTP (Cross-linked C-terminal telopeptide of Collagen alpha-1 (I) chain) ELISA Kit
Supplier:  Biomatik Corporation EKF57302
Encompass_Preferred
Human ICTP (Cross-linked C-terminal telopeptide of Collagen alpha-1 (I) chain) ELISA Kit for detecting ICTP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02452; P08123. Target Synonyms: ICTP; Cross-linked C-terminal telopeptide of Collagen alpha-1; I chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7917