Learn More
Biomatik Corporation Human C1QTNF1 (Complement C1q tumor necrosis factor-related protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57358
Encompass_Preferred
Human C1QTNF1 (Complement C1q tumor necrosis factor-related protein 1) ELISA Kit for detecting C1QTNF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9BXJ1. Target Synonyms: C1QTNF1; Complement C1q tumor necrosis factor-related protein 1; G protein-coupled receptor-interacting protein; GIP; UNQ310; PRO353; CTRP1; C1q and tumor necrosis factor related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7925
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'