Learn More
Biomatik Corporation Human WNT3A (Protein Wnt-3a) ELISA Kit
Supplier:  Biomatik Corporation EKF57462
Encompass_Preferred
Human WNT3A (Protein Wnt-3a) ELISA Kit for detecting WNT3A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P56704. Target Synonyms: WNT3A; MGC119418; MGC119419; MGC119420; protein Wnt-3a; wingless-type MMTV integRation site family; member 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7949