Learn More
Biomatik Corporation Mouse Lectin Galactoside Binding, Soluble 3 Binding Protein (LGALS3BP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05574
Encompass_Preferred
Mouse Lectin Galactoside Binding, Soluble 3 Binding Protein (LGALS3BP) ELISA Kit for detecting LGALS3BP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07797. Gene ID: 19039. Target Synonyms: MAC-2-BP; LGALS3-BP; Galectin-3-Binding Protein; L3 Antigen; Mac-2-Binding Protein; Tumor-associated antigen 90K; Serum Protein 90K; Basement membrane autoantigen p105. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3111