Learn More
Biomatik Corporation Rat MMP-13 (Matrix Metalloproteinase 13) ELISA Kit
Supplier:  Biomatik Corporation EKF57943
Encompass_Preferred
Rat MMP-13 (Matrix Metalloproteinase 13) ELISA Kit for detecting MMP-13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23097. Target Synonyms: MMP-13; CLG3; MANDP1; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8001