Learn More
Biomatik Corporation Mouse Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05577
Encompass_Preferred
Mouse Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit for detecting LOX1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQ09. Gene ID: 108078. Target Synonyms: OLR1; CLEC8A; SCARE1; Oxidized low-density lipoprotein receptor 1; C-type lectin domain family 8 member A; Lectin-like oxidized LDL receptor 1; Lectin-type oxidized LDL receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3113