Learn More
Biomatik Corporation Human Left/Right Determination Factor 1 (LEFTY1) ELISA Kit, 96 tests
Catalog No. 501503114
Encompass_Preferred
The Human Left/Right Determination Factor 1 (LEFTY1) ELISA Kit allows for quantitative determination of Left/Right Determination Factor 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.781-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75610. Cross Activity: No significant cross-reactivity or interference between Left/Right Determination Factor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3114 Supplier Biomatik Corporation Supplier No. EKU05581
The Human Left/Right Determination Factor 1 (LEFTY1) ELISA Kit allows for quantitative determination of Left/Right Determination Factor 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.781-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75610. Cross Activity: No significant cross-reactivity or interference between Left/Right Determination Factor 1 and analogues was observed. Restriction: For Research Use Only.