Learn More
Biomatik Corporation ALD (Aldosterone) ELISA Kit
Supplier:  Biomatik Corporation EKF58042
Encompass_Preferred
ALD (Aldosterone) ELISA Kit for detecting ALD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: ALD; Aldosterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8020