Learn More
Biomatik Corporation Human PYY (Peptide YY) ELISA Kit
Supplier:  Biomatik Corporation EKF58111
Encompass_Preferred
Human PYY (Peptide YY) ELISA Kit for detecting PYY in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P10082. Target Synonyms: PYY; Peptide YY; PYY-I; Peptide tyrosine tyrosine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8038