Learn More
Biomatik Corporation Horse Leptin (LEP) ELISA Kit, 96 tests
Catalog No. 501503116
Encompass_Preferred
The Horse Leptin (LEP) ELISA Kit allows for quantitative determination of Leptin in Serum, Plasma and other biological fluids. Species Reactivity: Equus caballus; Equine (Horse). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.223 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9TU09. Cross Activity: No significant cross-reactivity or interference between Leptin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3116 Supplier Biomatik Corporation Supplier No. EKU05591
The Horse Leptin (LEP) ELISA Kit allows for quantitative determination of Leptin in Serum, Plasma and other biological fluids. Species Reactivity: Equus caballus; Equine (Horse). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.223 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9TU09. Cross Activity: No significant cross-reactivity or interference between Leptin and analogues was observed. Restriction: For Research Use Only.