Learn More
Biomatik Corporation Mouse SOD1 (Superoxide Dismutase [Cu-Zn]) ELISA Kit
Supplier:  Biomatik Corporation EKF58138
Encompass_Preferred
Mouse SOD1 (Superoxide Dismutase [Cu-Zn]) ELISA Kit for detecting SOD1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml (10000iu/mg). Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08228. Target Synonyms: SOD1; Cu-Zn SOD; Ipo1; IPOA; SOD; cytosolic; SOD; Soluble; ALS; ALS1; amyotrophic lateral sclerosis 1; adult; Cu; Zn superoxide dismutase; hSod1; indophenoloxidase A; IPOA; SOD; Superoxide dismutase 1; superoxide dismutase 1; soluble; superoxide dismutase; cystolic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8044