Learn More
Biomatik Corporation Human Leptin (LEP) ELISA Kit, 96 tests
Catalog No. 501503117
Encompass_Preferred
The Human Leptin (LEP) ELISA Kit allows for quantitative determination of Leptin in Serum, Plasma, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41159. Cross Activity: No significant cross-reactivity or interference between Leptin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3117 Supplier Biomatik Corporation Supplier No. EKU05593
The Human Leptin (LEP) ELISA Kit allows for quantitative determination of Leptin in Serum, Plasma, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41159. Cross Activity: No significant cross-reactivity or interference between Leptin and analogues was observed. Restriction: For Research Use Only.