Learn More
Biomatik Corporation IP3 (Inositol Triphosphate) ELISA Kit
Supplier:  Biomatik Corporation EKF58173
Encompass_Preferred
IP3 (Inositol Triphosphate) ELISA Kit for detecting IP3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: IP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8055