Learn More
Biomatik Corporation Mouse NFkB (Nuclear Factor Kappa B) ELISA Kit
Supplier:  Biomatik Corporation EKF58215
Encompass_Preferred
Mouse NFkB (Nuclear Factor Kappa B) ELISA Kit for detecting NFkB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9WTK5; P25799; Q04863; Q04207. Target Synonyms: NFkB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8065