Learn More
Biomatik Corporation Rat Leptin (LEP) ELISA Kit, 96 tests
Catalog No. 501503120
Encompass_Preferred
Rat Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50596. Gene ID: 25608. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3120 Supplier Biomatik Corporation Supplier No. EKU05596
Rat Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50596. Gene ID: 25608. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.