Learn More
Biomatik Corporation Human AQP-3 (Aquaporin-3) ELISA Kit
Supplier:  Biomatik Corporation EKF58300
Encompass_Preferred
Human AQP-3 (Aquaporin-3) ELISA Kit for detecting AQP-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92482. Target Synonyms: AQP-3; Aquaporin-3; AQP3; GIL; Aquaglyceroporin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8091