Learn More
Biomatik Corporation Rabbit Leptin (LEP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05597
Encompass_Preferred
Rabbit Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B0KZL0. Gene ID: 100008747. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3121