Learn More
Biomatik Corporation Human HYAL2 (Hyaluronidase-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58341
Encompass_Preferred
Human HYAL2 (Hyaluronidase-2) ELISA Kit for detecting HYAL2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q12891. Target Synonyms: HYAL2; Hyaluronidase-2; Hyaluronoglucosaminidase-2; Lung carcinoma protein 2; LuCa-2; LUCA2; Hyal-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8102