Learn More
Biomatik Corporation Human Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05603
Encompass_Preferred
The Human Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) ELISA Kit allows for quantitative determination of Leucine Rich Alpha-2-Glycoprotein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.37 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02750. Cross Activity: No significant cross-reactivity or interference between Leucine Rich Alpha-2-Glycoprotein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3122