Learn More
Biomatik Corporation Mouse Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) ELISA Kit, 96 tests
Catalog No. 501503123
Encompass_Preferred
Mouse Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) ELISA Kit for detecting LRG1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91XL1. Gene ID: 76905. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3123 Supplier Biomatik Corporation Supplier No. EKU05604
Mouse Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) ELISA Kit for detecting LRG1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91XL1. Gene ID: 76905. Restrictions: For Research Use Only. Not for use in diagnostic procedures.