Learn More
Biomatik Corporation Human Leucine Rich Repeat In FLII Interacting Protein 1 (LRRFIP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05609
Encompass_Preferred
The Human Leucine Rich Repeat In FLII Interacting Protein 1 (LRRFIP1) ELISA Kit allows for quantitative determination of Leucine Rich Repeat In FLII Interacting Protein 1 in Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q32MZ4. Cross Activity: No significant cross-reactivity or interference between Leucine Rich Repeat In FLII Interacting Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3124