Learn More
Biomatik Corporation AA (Arachidonic Acid) ELISA Kit
Supplier:  Biomatik Corporation EKF58511
Encompass_Preferred
AA (Arachidonic Acid) ELISA Kit for detecting AA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100ug/ml. Sensitivity: 0.938ug/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: AA; Arachidonic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8144