Learn More
Biomatik Corporation Human PIF/DCD (Proteolysis Inducing Factor/Dermcidin) ELISA Kit
Supplier:  Biomatik Corporation EKF58737
Encompass_Preferred
Human PIF/DCD (Proteolysis Inducing Factor/Dermcidin) ELISA Kit for detecting PIF; DCD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P81605. Target Synonyms: AIDD; DCD-1; DSEP; HCAP; MGC71930; PIF; diffusible survival; evasion peptide; preproteolysin; proteolysis inducing factor; survival promoting peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8180