Learn More
Biomatik Corporation Mouse Leukocyte Cell Derived Chemotaxin 2 (LECT2) ELISA Kit, 96 tests
Catalog No. 501503128
Encompass_Preferred
Mouse Leukocyte Cell Derived Chemotaxin 2 (LECT2) ELISA Kit for detecting LECT2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88803. Gene ID: 16841. Target Synonyms: Chm-II; Chm2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3128 Supplier Biomatik Corporation Supplier No. EKU05624
Mouse Leukocyte Cell Derived Chemotaxin 2 (LECT2) ELISA Kit for detecting LECT2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88803. Gene ID: 16841. Target Synonyms: Chm-II; Chm2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.