Learn More
Biomatik Corporation Human OCLN (Occludin) ELISA Kit
Supplier:  Biomatik Corporation EKF58865
Encompass_Preferred
Human OCLN (Occludin) ELISA Kit for detecting OCLN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q16625. Target Synonyms: OCLN; Occludin; BLCPMG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8196