Learn More
Biomatik Corporation Human IFN-α (Interferon Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF58911
Encompass_Preferred
Human IFN-α (Interferon Alpha) ELISA Kit for detecting IFN-α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01562. Target Synonyms: IFN-α; Interferon Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8207