Learn More
Biomatik Corporation Human Lipase, Bile Salt Dependent (BSDL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05639
Encompass_Preferred
The Human Lipase, Bile Salt Dependent (BSDL) ELISA Kit allows for quantitative determination of Lipase, Bile Salt Dependent in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19835. Cross Activity: No significant cross-reactivity or interference between Lipase, Bile Salt Dependent and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3133