Learn More
Biomatik Corporation Human Lipase, Endothelial (LIPG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05640
Encompass_Preferred
The Human Lipase, Endothelial (LIPG) ELISA Kit allows for quantitative determination of Lipase, Endothelial in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y5X9. Cross Activity: No significant cross-reactivity or interference between Lipase, Endothelial and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3134