Learn More
Biomatik Corporation Mouse PRF1 (Perforin 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58989
Encompass_Preferred
Mouse PRF1 (Perforin 1) ELISA Kit for detecting PRF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10820. Target Synonyms: PRF1; Perforin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8223