Learn More
Biomatik Corporation Rat Lipase, Hormone Sensitive (LIPE) ELISA Kit, 96 tests
Catalog No. 501503137
Encompass_Preferred
Rat Lipase, Hormone Sensitive (LIPE) ELISA Kit for detecting LIPE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15304. Gene ID: 25330. Target Synonyms: HSL; LHS; Hormone Sensitive Lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3137 Supplier Biomatik Corporation Supplier No. EKU05647
Rat Lipase, Hormone Sensitive (LIPE) ELISA Kit for detecting LIPE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15304. Gene ID: 25330. Target Synonyms: HSL; LHS; Hormone Sensitive Lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.