Learn More
Biomatik Corporation Human PIK3CA (Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform) ELISA Kit
Supplier:  Biomatik Corporation EKF59031
Encompass_Preferred
Human PIK3CA (Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform) ELISA Kit for detecting PIK3CA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P42336. Target Synonyms: PIK3CA; Phosphatidylinositol-4; 5-bisphosphate 3-kinase catalytic subunit alpha isoform; PI3-kinase subunit alpha; PI3K-alpha; PI3Kalpha; PtdIns-3-kinase subunit alpha; Phosphatidylinositol-4; 5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; PtdIns-3-kinase subunit p110-alpha; p110alpha; Phosphoinositide-3-kinase catalytic alpha polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8228