Learn More
Biomatik Corporation Rat Arg1 (Arginase-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59033
Encompass_Preferred
Rat Arg1 (Arginase-1) ELISA Kit for detecting Arg1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.5-160ng/ml. Sensitivity: 1.5ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07824. Target Synonyms: Arg1; arginase; liver; arginase-1; Liver-type arginase; Type I arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8230