Learn More
Biomatik Corporation Mouse Lipase, Lipoprotein (LIPD) ELISA Kit, 96 tests
Catalog No. 501503139
Encompass_Preferred
Mouse Lipoprotein lipase (LPL) ELISA Kit for detecting LPL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11152. Gene ID: 16956. Target Synonyms: LIPD; Lipase, Lipoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3139 Supplier Biomatik Corporation Supplier No. EKU05649
Mouse Lipoprotein lipase (LPL) ELISA Kit for detecting LPL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11152. Gene ID: 16956. Target Synonyms: LIPD; Lipase, Lipoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.