Learn More
Biomatik Corporation Mouse CAMP (Cathelicidin Antimicrobial Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF59059
Encompass_Preferred
Mouse CAMP (Cathelicidin Antimicrobial Peptide) ELISA Kit for detecting CAMP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51437. Target Synonyms: CAMP; CAP 18; CAP18; CRAMP; FALL 39; FALL39; hCAP 18; HSD26; LL37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8234