Learn More
Biomatik Corporation Rat Lipase, Lipoprotein (LIPD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05650
Encompass_Preferred
Rat Lipoprotein lipase (LPL) ELISA Kit for detecting LPL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.41ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06000. Gene ID: 24539. Target Synonyms: LIPD; Lipase, Lipoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3140