Learn More
Biomatik Corporation Mouse Lipase, Pancreatic (PL) ELISA Kit, 96 tests
Catalog No. 501503141
Encompass_Preferred
Mouse Pancreatic Lipase (PL) ELISA Kit for detecting PL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P8U6. Gene ID: 69060. Target Synonyms: PNLIP; PTL; Pancreatic Lipase; Pancreatic triacylglycerol lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3141 Supplier Biomatik Corporation Supplier No. EKU05652
Mouse Pancreatic Lipase (PL) ELISA Kit for detecting PL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P8U6. Gene ID: 69060. Target Synonyms: PNLIP; PTL; Pancreatic Lipase; Pancreatic triacylglycerol lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.