Learn More
Biomatik Corporation Mouse MuRF1 (muscle-specific RING-finger protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59097
Encompass_Preferred
Mouse MuRF1 (muscle-specific RING-finger protein 1) ELISA Kit for detecting MuRF1 in cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q38HM4. Target Synonyms: MuRF1; IRF; RNF28; SMRZ; TRIM63; IRFMURF2; Iris RING finger protein; MuRF1; MURF1; MuRF-1; muscle specific ring finger protein 2; Muscle-specific RING finger protein 1; RING finger protein 28FLJ32380; RNF28E3 ubiquitin-protein ligase TRIM63; SMRZMURF-1; Striated muscle RING zinc finger protein; tripartite motif containing 63; tripartite motif-containing 63; Tripartite motif-containing protein 63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8238