Learn More
Biomatik Corporation Human Lipocalin 1 (LCN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05656
Encompass_Preferred
The Human Lipocalin 1 (LCN1) ELISA Kit allows for quantitative determination of Lipocalin 1 in Serum, Plasma, Saliva, tears and other biological fluids and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.62 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P31025. Cross Activity: No significant cross-reactivity or interference between Lipocalin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3143