Learn More
Biomatik Corporation Cattle Lipopolysaccharide Binding Protein (LBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05661
Encompass_Preferred
The Cattle Lipopolysaccharide Binding Protein (LBP) ELISA Kit allows for quantitative determination of Lipopolysaccharide Binding Protein in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.287 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q2TBI0. Cross Activity: No significant cross-reactivity or interference between Lipopolysaccharide Binding Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3144