Learn More
Biomatik Corporation Human Carbamylated Albumin ELISA Kit
Supplier:  Biomatik Corporation EKF59175
Encompass_Preferred
Human Carbamylated Albumin ELISA Kit for detecting Carbamylated Albumin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02768. Target Synonyms: Carbamylated Albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8248