Learn More
Biomatik Corporation Mouse Lipopolysaccharide Binding Protein (LBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05663
Encompass_Preferred
Mouse Lipopolysaccharide Binding Protein (LBP) ELISA Kit for detecting LBP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61805. Gene ID: 16803. Target Synonyms: LPS-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3146