Learn More
Biomatik Corporation Human ARC (Activity-regulated cytoskeleton-associated protein) ELISA Kit
Supplier:  Biomatik Corporation EKF59199
Encompass_Preferred
Human ARC (Activity-regulated cytoskeleton-associated protein) ELISA Kit for detecting ARC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7LC44. Target Synonyms: ARC; ARG3.1; mArc; Activity-regulated gene 3.1 protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8250