Learn More
Biomatik Corporation Lipoxin A4 (LXA4) ELISA Kit, 96 tests
Catalog No. 501503149
Encompass_Preferred
Lipoxin A4 (LXA4) ELISA Kit for detecting LXA4 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 493.8-40,000pg/ml. Sensitivity: 166.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: LX-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3149 Supplier Biomatik Corporation Supplier No. EKU05667
Lipoxin A4 (LXA4) ELISA Kit for detecting LXA4 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 493.8-40,000pg/ml. Sensitivity: 166.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: LX-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.