Learn More
Biomatik Corporation Chicken SAA (serum amyloid A) ELISA Kit
Supplier:  Biomatik Corporation EKF59292
Encompass_Preferred
Chicken SAA (serum amyloid A) ELISA Kit for detecting SAA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: F1NW65. Target Synonyms: Serum amyloid A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8260